Product Details
- SNP ID
-
rs832195
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:63878936 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAGTGCCTTCTATTGGCCAGAC[C/T]CACCTGGAAGCTGGCCAGCAGGTGA
- Phenotype
-
MIM: 607640
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN7
PubMed Links
Gene Details
- Gene
- ATXN7
- Gene Name
- ataxin 7
View Full Product Details