Product Details
- SNP ID
-
rs789295
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:154341087 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAAGTTAAAGGGTTATATTGAAAA[C/T]TTTCCATCCTGAGAAAACATTTTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPR149
PubMed Links
Gene Details
- Gene
- GPR149
- Gene Name
- G protein-coupled receptor 149
View Full Product Details