Product Details
- SNP ID
-
rs701265
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:152836568 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGAGGAGAAAATCGATTTACCTGGT[A/G]ATCATTGTACTGACTGTTTTTGCTG
- Phenotype
-
MIM: 601167
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
P2RY1
PubMed Links
Gene Details
- Gene
- P2RY1
- Gene Name
- purinergic receptor P2Y1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002563.4 |
1876 |
Silent Mutation |
GTA,GTG |
V262V |
NP_002554.1 |
View Full Product Details