Product Details

SNP ID
rs7032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:30709719 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCACAATTTATAAAGTGCCAGAT[A/T]AGTGCTAATTGTCATTCAGCTTGAT
Phenotype
MIM: 610375
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPRIN2 PubMed Links

Gene Details

Gene
CAPRIN2
Gene Name
caprin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002259.2 3494 UTR 3 NP_001002259.1
NM_001206856.2 3494 UTR 3 NP_001193785.1
NM_001319842.1 3494 UTR 3 NP_001306771.1
NM_001319843.1 3494 UTR 3 NP_001306772.1
NM_001319844.1 3494 UTR 3 NP_001306773.1
NM_001319845.1 3494 UTR 3 NP_001306774.1
NM_001319846.1 3494 UTR 3 NP_001306775.1
NM_023925.4 3494 UTR 3 NP_076414.2
NM_032156.4 3494 UTR 3 NP_115532.3
XM_006719138.3 3494 UTR 3 XP_006719201.1
XM_006719139.3 3494 UTR 3 XP_006719202.1
XM_006719140.3 3494 UTR 3 XP_006719203.1
XM_006719142.3 3494 UTR 3 XP_006719205.1
XM_006719144.3 3494 UTR 3 XP_006719207.1
XM_006719145.3 3494 UTR 3 XP_006719208.1
XM_006719146.3 3494 Intron XP_006719209.1
XM_006719147.3 3494 UTR 3 XP_006719210.1
XM_017019851.1 3494 UTR 3 XP_016875340.1
XM_017019852.1 3494 UTR 3 XP_016875341.1
XM_017019853.1 3494 UTR 3 XP_016875342.1
XM_017019854.1 3494 UTR 3 XP_016875343.1
XM_017019855.1 3494 UTR 3 XP_016875344.1
XM_017019856.1 3494 UTR 3 XP_016875345.1
XM_017019857.1 3494 UTR 3 XP_016875346.1
XM_017019858.1 3494 UTR 3 XP_016875347.1
XM_017019859.1 3494 UTR 3 XP_016875348.1
XM_017019860.1 3494 UTR 3 XP_016875349.1
XM_017019861.1 3494 UTR 3 XP_016875350.1
XM_017019862.1 3494 UTR 3 XP_016875351.1
XM_017019863.1 3494 Intron XP_016875352.1
XM_017019864.1 3494 Intron XP_016875353.1
XM_017019865.1 3494 UTR 3 XP_016875354.1
XM_017019866.1 3494 UTR 3 XP_016875355.1
XM_017019867.1 3494 UTR 3 XP_016875356.1
XM_017019868.1 3494 UTR 3 XP_016875357.1
XM_017019869.1 3494 UTR 3 XP_016875358.1
XM_017019870.1 3494 UTR 3 XP_016875359.1
XM_017019871.1 3494 UTR 3 XP_016875360.1
XM_017019872.1 3494 UTR 3 XP_016875361.1
XM_017019873.1 3494 UTR 3 XP_016875362.1
XM_017019874.1 3494 UTR 3 XP_016875363.1
XM_017019875.1 3494 UTR 3 XP_016875364.1
XM_017019876.1 3494 UTR 3 XP_016875365.1
XM_017019877.1 3494 UTR 3 XP_016875366.1
XM_017019878.1 3494 UTR 3 XP_016875367.1
XM_017019879.1 3494 UTR 3 XP_016875368.1
XM_017019880.1 3494 UTR 3 XP_016875369.1
XM_017019881.1 3494 UTR 3 XP_016875370.1
XM_017019882.1 3494 UTR 3 XP_016875371.1
XM_017019883.1 3494 UTR 3 XP_016875372.1

View Full Product Details