Product Details

SNP ID
rs1229430
Assay Type
Validated
NCBI dbSNP Submissions
76
Location
Chr.1:167544181 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GCCCGTGCCTCTCCTGCATGCACAG[C/T]AGCAAAACAAGCGCCAGGACTGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREG1 PubMed Links

Gene Details

Gene
CREG1
Gene Name
cellular repressor of E1A stimulated genes 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003851.2 Intron NP_003842.1

View Full Product Details