Product Details

SNP ID
rs1420564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:47164429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAAGGCATGCCTTCAAAGTTCA[A/G]AAAATGACAGCCTTAGTTTTCATTT
Phenotype
MIM: 611803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITFG1 PubMed Links

Gene Details

Gene
ITFG1
Gene Name
integrin alpha FG-GAP repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305002.1 Intron NP_001291931.1
NM_030790.4 Intron NP_110417.2
Gene
ITFG1-AS1
Gene Name
ITFG1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details