Product Details

SNP ID
rs811734
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:46804243 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCAACCTGCTCCAGTATGAAATG[A/G]TTGCTCTCTAGGCCTACTGCAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf87 PubMed Links

Gene Details

Gene
C16orf87
Gene Name
chromosome 16 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001436.2 Intron NP_001001436.1

View Full Product Details