Product Details

SNP ID
rs645196
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:170663925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGACTTTTTAATATTGTATTTTA[C/G]TGTGGATTATCTCTTTGGACCGCGC
Phenotype
MIM: 167420
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRRX1 PubMed Links

Gene Details

Gene
PRRX1
Gene Name
paired related homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006902.4 Intron NP_008833.1
NM_022716.3 Intron NP_073207.1
XM_006711388.3 Intron XP_006711451.1

View Full Product Details