Product Details

SNP ID
rs1293768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112986696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTGGCCGCAGGTGGGACAGTTT[G/T]ACCCCCATTCCCCCAGATGCGCACA
Phenotype
MIM: 603350
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OAS2 PubMed Links

Gene Details

Gene
OAS2
Gene Name
2'-5'-oligoadenylate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032731.1 Intron NP_001027903.1
NM_002535.2 Intron NP_002526.2
NM_016817.2 Intron NP_058197.2

View Full Product Details