Product Details
- SNP ID
-
rs920224
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:109389105 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGTAGGTCCCGGGACTAGATCTAG[A/G]GTTTCAGACAATGTCTGGCAACTCC
- Phenotype
-
MIM: 614636
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1H
PubMed Links
Gene Details
- Gene
- MYO1H
- Gene Name
- myosin IH
View Full Product Details