Product Details

SNP ID
rs1713419
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:20366389 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGACTAGGATGATGCCATGCCAT[A/G]GTTCGCTGCAGGATTTCTGGCAAGG
Phenotype
MIM: 607725 MIM: 601686
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PARP2 PubMed Links
Additional Information
For this assay, SNP(s) [rs77605789] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PARP2
Gene Name
poly(ADP-ribose) polymerase 2
There are no transcripts associated with this gene.

Gene
TEP1
Gene Name
telomerase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319035.1 8457 UTR 3 NP_001305964.1
NM_007110.4 8457 UTR 3 NP_009041.2
XM_005268027.4 8457 UTR 3 XP_005268084.1
XM_011537110.2 8457 UTR 3 XP_011535412.1

View Full Product Details