Product Details

SNP ID
rs1050640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6748082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGGCAGAGAAGGGTGTAGGGAA[A/G]ATTCTTTTGATGTGTGTGGGCAGGA
Phenotype
MIM: 601401
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLF2 PubMed Links

Gene Details

Gene
MLF2
Gene Name
myeloid leukemia factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005439.2 1301 UTR 3 NP_005430.1

View Full Product Details