Product Details

SNP ID
rs1713452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20426052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTAGTCTATCTTAAATAGAGA[A/T]ACAGAAAAAGATCTTACTAAAATAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL33 PubMed Links

Gene Details

Gene
KLHL33
Gene Name
kelch like family member 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109997.2 5621 Intron NP_001103467.2
XM_005267347.4 5621 UTR 3 XP_005267404.1
XM_011536450.2 5621 Intron XP_011534752.1

View Full Product Details