Product Details

SNP ID
rs1263645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22548380 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAACTCAGGGTTGAGAAAACAGC[C/T]ACCTTCAGGACAAAAGTCAGGGAAG
Phenotype
MIM: 186880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370399 PubMed Links

Gene Details

Gene
LOC105370399
Gene Name
uncharacterized LOC105370399
There are no transcripts associated with this gene.

Gene
TRAC
Gene Name
T-cell receptor alpha constant
There are no transcripts associated with this gene.

View Full Product Details