Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014631.2 | 13581 | UTR 3 | NP_055446.2 | ||
XM_005270294.4 | 13581 | UTR 3 | XP_005270351.1 | ||
XM_005270295.4 | 13581 | UTR 3 | XP_005270352.1 | ||
XM_005270297.3 | 13581 | UTR 3 | XP_005270354.1 | ||
XM_005270298.4 | 13581 | UTR 3 | XP_005270355.1 | ||
XM_006718079.3 | 13581 | UTR 3 | XP_006718142.1 | ||
XM_006718080.3 | 13581 | UTR 3 | XP_006718143.1 | ||
XM_011540385.2 | 13581 | UTR 3 | XP_011538687.1 | ||
XM_011540386.2 | 13581 | UTR 3 | XP_011538688.1 | ||
XM_011540387.2 | 13581 | UTR 3 | XP_011538689.1 | ||
XM_017016969.1 | 13581 | UTR 3 | XP_016872458.1 | ||
XM_017016970.1 | 13581 | UTR 3 | XP_016872459.1 |