Product Details

SNP ID
rs859311
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:4737102 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGTCCCTGAAATGGCTCTCAAAGA[C/T]GCTCAGACCAATAGCCCTAGGTGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS3 PubMed Links

Gene Details

Gene
ANKS3
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
There are no transcripts associated with this gene.

Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 Intron NP_631909.2
XM_005255144.3 Intron XP_005255201.1
XM_017022975.1 Intron XP_016878464.1
XM_017022976.1 Intron XP_016878465.1
Gene
ZNF500
Gene Name
zinc finger protein 500
There are no transcripts associated with this gene.

View Full Product Details