Product Details

SNP ID
rs1270299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18842526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTCTGTAGCTTTGGAATTTTT[A/C]AAAATAAGAGGAAATATCAGGGCAG
Phenotype
MIM: 601430
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UPF1 PubMed Links

Gene Details

Gene
UPF1
Gene Name
UPF1, RNA helicase and ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297549.1 Intron NP_001284478.1
NM_002911.3 Intron NP_002902.2
XM_017027105.1 Intron XP_016882594.1
XM_017027106.1 Intron XP_016882595.1

View Full Product Details