Product Details
- SNP ID
-
rs1320051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:48188238 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTAAGGTGATTAAAAAAAAAGAA[A/G]AGATATAAAAGGCTTAAGGTCTTAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTXN2
PubMed Links
Gene Details
- Gene
- CTXN2
- Gene Name
- cortexin 2
- Gene
- MYEF2
- Gene Name
- myelin expression factor 2
There are no transcripts associated with this gene.
View Full Product Details