Product Details
- SNP ID
-
rs926263
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:167786691 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AACAGGAAAAGGCAGTTAACAAATT[C/G]ATACATTCTCCTCTTCCTACTTCTA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01558
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139592860] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01558
- Gene Name
- long intergenic non-protein coding RNA 1558
There are no transcripts associated with this gene.
- Gene
- LOC441179
- Gene Name
- uncharacterized LOC441179
There are no transcripts associated with this gene.
View Full Product Details