Product Details

SNP ID
rs1062897
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:93346032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCACGGCGGCAGCCACTGCGAC[A/T]CCCACTGTGCCTGGCTCTGTCCATA
Phenotype
MIM: 601482
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC18-AS1 PubMed Links

Gene Details

Gene
CCDC18-AS1
Gene Name
CCDC18 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DR1
Gene Name
down-regulator of transcription 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001938.2 112 UTR 5 NP_001929.1

View Full Product Details