Product Details

SNP ID
rs1617629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31069021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCAGCCCCCCGGCAGGATTCC[G/A]AGGTCTGGTGTCCTCCTTTCACCAT
Phenotype
MIM: 125645
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DSC2 PubMed Links

Gene Details

Gene
DSC2
Gene Name
desmocollin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004949.4 2842 Missense Mutation TCG,TTG S794L NP_004940.1
NM_024422.4 2842 Missense Mutation TCG,TTG S794L NP_077740.1
XM_005258206.4 2842 Missense Mutation TCG,TTG S651L XP_005258263.1

View Full Product Details