Product Details

SNP ID
rs1804649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102631893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTGGGTAAACAGCCTCCCCTG[A/G]AAGTGTTCAGTGTACAACTCTTCAC
Phenotype
MIM: 609489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MANBA PubMed Links

Gene Details

Gene
MANBA
Gene Name
mannosidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005908.3 2039 UTR 3 NP_005899.3
XM_011531966.2 2039 UTR 3 XP_011530268.1
XM_017008203.1 2039 UTR 3 XP_016863692.1
XM_017008204.1 2039 UTR 3 XP_016863693.1
XM_017008205.1 2039 UTR 3 XP_016863694.1

View Full Product Details