Product Details

SNP ID
rs1025910
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:52344970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTATGATAGCTCTAATTACATAA[C/G]GGCACTTGCAATTTCCCTAGAGAAT
Phenotype
MIM: 120470
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DCC PubMed Links

Gene Details

Gene
DCC
Gene Name
DCC netrin 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005215.3 Intron NP_005206.2
XM_011525843.1 Intron XP_011524145.1
XM_011525844.2 Intron XP_011524146.1
XM_017025568.1 Intron XP_016881057.1
XM_017025569.1 Intron XP_016881058.1
XM_017025570.1 Intron XP_016881059.1

View Full Product Details