Product Details
- SNP ID
-
rs995593
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:47709098 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTGGGTTACTGGGTAGGTCACTA[A/G]ACAGGCAGGATTGGCTCTGGAATGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CXXC1P1
PubMed Links
Gene Details
- Gene
- CXXC1P1
- Gene Name
- CXXC finger protein 1 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details