Product Details

SNP ID
rs1059579
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:47639001 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGCTGCTGCCCCTGCAGGAGCTGCA[C/T]TGGGGAGCACCACAGCAGGCCGAGG
Phenotype
MIM: 300383 MIM: 311040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFP PubMed Links

Gene Details

Gene
CFP
Gene Name
complement factor properdin
There are no transcripts associated with this gene.

Gene
ELK1
Gene Name
ELK1, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114123.2 764 Missense Mutation AAT,AGT N183S NP_001107595.1
NM_001257168.1 764 Intron NP_001244097.1
NM_005229.4 764 Missense Mutation AAT,AGT N183S NP_005220.2
XM_017029339.1 764 Missense Mutation AAT,AGT N183S XP_016884828.1

View Full Product Details