Product Details
- SNP ID
-
rs3915966
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31054497 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGGACTCAGACAGGGCCAGTCC[C/T]GAGCGAAGGTACGGAATAGGTGAGT
- Phenotype
-
MIM: 613918
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HCG22
PubMed Links
Gene Details
- Gene
- HCG22
- Gene Name
- HLA complex group 22
There are no transcripts associated with this gene.
View Full Product Details