Product Details

SNP ID
rs3906277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30909456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGAGCTCCCATCCTTGGCTAA[A/G]AACTGGGTGATGCGGATGCTCTTTC
Phenotype
MIM: 600408 MIM: 601760 MIM: 612802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDR1 PubMed Links

Gene Details

Gene
DDR1
Gene Name
discoidin domain receptor tyrosine kinase 1
There are no transcripts associated with this gene.

Gene
GTF2H4
Gene Name
general transcription factor IIH subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001517.4 366 Silent Mutation AAA,AAG K53K NP_001508.1
Gene
VARS2
Gene Name
valyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details