Product Details

SNP ID
rs1267068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161155624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTAGATGTTATTCTATTCAGCT[G/T]TTGAAGTTTACTGCTGAAGACAAGT
Phenotype
MIM: 603893
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TANK PubMed Links

Gene Details

Gene
TANK
Gene Name
TRAF family member associated NFKB activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199135.1 Intron NP_001186064.1
NM_004180.2 Intron NP_004171.2
NM_133484.1 Intron NP_597841.1
XM_005246207.3 Intron XP_005246264.2
XM_017003093.1 Intron XP_016858582.1
XM_017003094.1 Intron XP_016858583.1
XM_017003095.1 Intron XP_016858584.1
XM_017003096.1 Intron XP_016858585.1
XM_017003097.1 Intron XP_016858586.1

View Full Product Details