Product Details

SNP ID
rs9713
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:161308497 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTGATTCATCGCCGGTTTGCAGAC[A/T]GAGCCGCGTCGGGTGTGCGCCGCTG
Phenotype
MIM: 607173
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PSMD14 PubMed Links

Gene Details

Gene
PSMD14
Gene Name
proteasome 26S subunit, non-ATPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005805.5 223 UTR 5 NP_005796.1

View Full Product Details