Product Details
- SNP ID
-
rs1055889
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:89637750 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATGATTTTGTTCTCACCTCTTCGG[G/T]GGCACTTTTGTTTCTTCTGAATTCT
- Phenotype
-
MIM: 610583
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115085953] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD6
- Gene Name
- ankyrin repeat domain 6
There are no transcripts associated with this gene.
- Gene
- LOC101929057
- Gene Name
- uncharacterized LOC101929057
There are no transcripts associated with this gene.
- Gene
- LYRM2
- Gene Name
- LYR motif containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020466.4 |
217 |
Missense Mutation |
ACC,CCC |
T60P |
NP_065199.1 |
- Gene
- MDN1
- Gene Name
- midasin AAA ATPase 1
There are no transcripts associated with this gene.
View Full Product Details