Product Details

SNP ID
rs1046434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:88612650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCGTTTTTCAATTTCTCTGGC[G/T]ACAAAAATGGGCAACAGCGTTTTTT
Phenotype
MIM: 603512
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNGTT PubMed Links

Gene Details

Gene
RNGTT
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286426.1 2014 UTR 3 NP_001273355.1
NM_001286428.1 2014 UTR 3 NP_001273357.1
NM_003800.4 2014 UTR 3 NP_003791.3
XM_017011401.1 2014 UTR 3 XP_016866890.1
XM_017011402.1 2014 Intron XP_016866891.1

View Full Product Details