Product Details

SNP ID
rs927330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:135810618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGCATTCATGGTTGCTACTGCA[C/T]TGGAATAATGGTAACAACAGGCTGA
Phenotype
MIM: 604645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928326 PubMed Links

Gene Details

Gene
LOC101928326
Gene Name
uncharacterized LOC101928326
There are no transcripts associated with this gene.

Gene
PDE7B
Gene Name
phosphodiesterase 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018945.3 Intron NP_061818.1
XM_005266931.3 Intron XP_005266988.1
XM_011535755.2 Intron XP_011534057.1
XM_017010741.1 Intron XP_016866230.1

View Full Product Details