Product Details

SNP ID
rs1054345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56525849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAGGCGGCCAACTTCACTCTCCA[C/T]TAAAGAAAAAAAGAAATGCTTCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM209A PubMed Links

Gene Details

Gene
FAM209A
Gene Name
family with sequence similarity 209 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012971.3 375 Silent Mutation CTA,TTA L99L NP_001012989.2
Gene
FAM209B
Gene Name
family with sequence similarity 209 member B
There are no transcripts associated with this gene.

Gene
GCNT7
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080615.1 375 UTR 5 NP_542182.1
Gene
RTFDC1
Gene Name
replication termination factor 2 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details