Product Details

SNP ID
rs1323040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:154010889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTACATGCAATAAATGTAATTCT[A/G]TGAGAAGGACCAGCCCTTACATCCC
Phenotype
MIM: 600018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OPRM1 PubMed Links

Gene Details

Gene
OPRM1
Gene Name
opioid receptor mu 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000914.4 394 Intron NP_000905.3
NM_001008503.2 394 Intron NP_001008503.2
NM_001008504.3 394 Intron NP_001008504.2
NM_001008505.2 394 Intron NP_001008505.2
NM_001145279.3 394 UTR 5 NP_001138751.1
NM_001145280.3 394 UTR 5 NP_001138752.1
NM_001145281.2 394 Intron NP_001138753.1
NM_001145282.2 394 Intron NP_001138754.1
NM_001145283.2 394 Intron NP_001138755.1
NM_001145284.3 394 Intron NP_001138756.1
NM_001145285.2 394 Intron NP_001138757.1
NM_001145286.2 394 Intron NP_001138758.1
NM_001145287.2 394 Intron NP_001138759.1
NM_001285522.1 394 Intron NP_001272451.1
NM_001285523.1 394 Intron NP_001272452.1
NM_001285524.1 394 Intron NP_001272453.1
NM_001285526.1 394 Intron NP_001272455.1
NM_001285527.1 394 Intron NP_001272456.1
NM_001285528.1 394 Intron NP_001272457.1
XM_011535851.2 394 Intron XP_011534153.1
XM_011535853.2 394 Intron XP_011534155.1
XM_011535856.2 394 Intron XP_011534158.1
XM_011535862.2 394 Intron XP_011534164.1
XM_017010903.1 394 Intron XP_016866392.1
XM_017010904.1 394 Intron XP_016866393.1
XM_017010905.1 394 Intron XP_016866394.1
XM_017010906.1 394 Intron XP_016866395.1
XM_017010907.1 394 Intron XP_016866396.1

View Full Product Details