Product Details

SNP ID
rs61741656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1566478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAGCAGTAGGAGCCAGCGCTG[C/G]TTCTGGAAATCAGGGAAGAGGAGGA
Phenotype
MIM: 603889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SIRPB1 PubMed Links

Gene Details

Gene
SIRPB1
Gene Name
signal regulatory protein beta 1
There are no transcripts associated with this gene.

Gene
SIRPD
Gene Name
signal regulatory protein delta
There are no transcripts associated with this gene.

View Full Product Details