Product Details

SNP ID
rs1984296
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:61080016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTAGTGTTCCCCCGCCTCCCCC[C/G]CTCCCTCCGCGGGGGAGGGAGCGGG
Phenotype
MIM: 600727
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NFIA PubMed Links

Gene Details

Gene
NFIA
Gene Name
nuclear factor I A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134673.3 Intron NP_001128145.1
NM_001145511.1 Intron NP_001138983.1
NM_001145512.1 Intron NP_001138984.1
NM_005595.4 Intron NP_005586.1
XM_011541512.2 Intron XP_011539814.1
XM_011541514.2 Intron XP_011539816.1
XM_011541515.2 Intron XP_011539817.1
XM_017001362.1 Intron XP_016856851.1
XM_017001363.1 Intron XP_016856852.1

View Full Product Details