Product Details

SNP ID
rs10889499
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:64754200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAACAAAGATGCAAAATTAATGT[C/G]AAACAATTTGCCCAAGATCATTCAG
Phenotype
MIM: 609953
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAVER2 PubMed Links

Gene Details

Gene
RAVER2
Gene Name
ribonucleoprotein, PTB binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018211.3 Intron NP_060681.2
XM_006710738.3 Intron XP_006710801.2
XM_011541706.2 Intron XP_011540008.1

View Full Product Details