Product Details
- SNP ID
-
rs34364530
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:91681032 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATTTTAACAACAGCTTCAGCAT[C/G]AAACAAACAACAAAATGGCCTCTGC
- Phenotype
-
MIM: 600742
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TGFBR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79001261] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TGFBR3
- Gene Name
- transforming growth factor beta receptor 3
View Full Product Details