Product Details

SNP ID
rs12492777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40396114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTTGGTAACAGACCCTAGCCA[C/T]CATTGCAAGGGACAGGAAATTGTAA
Phenotype
MIM: 603161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENTPD3 PubMed Links

Gene Details

Gene
ENTPD3
Gene Name
ectonucleoside triphosphate diphosphohydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248.3 Intron NP_001239.2
NM_001291960.1 Intron NP_001278889.1
NM_001291961.1 Intron NP_001278890.1
XM_011534266.2 Intron XP_011532568.1
Gene
ENTPD3-AS1
Gene Name
ENTPD3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details