Product Details

SNP ID
rs12460465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34682640 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAACTGTCCTATTATTACTTACC[A/T]GACATCACATTCCCAGTCATTGTCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCGB1B2P PubMed Links

Gene Details

Gene
SCGB1B2P
Gene Name
secretoglobin family 1B member 2, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF302
Gene Name
zinc finger protein 302
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012320.2 Intron NP_001012320.1
NM_001289181.1 Intron NP_001276110.1
NM_001289182.1 Intron NP_001276111.1
NM_001289183.1 Intron NP_001276112.1
NM_001289184.1 Intron NP_001276113.1
NM_001289185.1 Intron NP_001276114.1
NM_001289186.1 Intron NP_001276115.1
NM_001289187.1 Intron NP_001276116.1
NM_001289188.1 Intron NP_001276117.1
NM_001289189.1 Intron NP_001276118.1
NM_001289190.1 Intron NP_001276119.1
NM_001289191.1 Intron NP_001276120.1
NM_001289192.1 Intron NP_001276121.1
NM_018443.3 Intron NP_060913.2
NM_018675.2 Intron NP_061145.2
XM_011527111.2 Intron XP_011525413.1
XM_017026978.1 Intron XP_016882467.1
XM_017026979.1 Intron XP_016882468.1
XM_017026980.1 Intron XP_016882469.1
XM_017026981.1 Intron XP_016882470.1
XM_017026982.1 Intron XP_016882471.1
XM_017026983.1 Intron XP_016882472.1
XM_017026984.1 Intron XP_016882473.1
XM_017026985.1 Intron XP_016882474.1
XM_017026986.1 Intron XP_016882475.1

View Full Product Details