Product Details

SNP ID
rs10911339
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:183472962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCGAGGGGTGGAGAGAAACGTC[C/T]GGGGAGGCGGCGGCGACCCCGAGTG
Phenotype
MIM: 610964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMG7 PubMed Links

Gene Details

Gene
SMG7
Gene Name
SMG7, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174061.1 Intron NP_001167532.1
NM_173156.2 Intron NP_775179.1
NM_201568.2 Intron NP_963862.1
NM_201569.2 Intron NP_963863.2
XM_005245648.3 Intron XP_005245705.1
XM_005245649.3 Intron XP_005245706.1
XM_005245652.3 Intron XP_005245709.1
XM_005245653.4 Intron XP_005245710.1
XM_006711676.3 Intron XP_006711739.1
XM_006711678.3 Intron XP_006711741.1
XM_011510205.2 Intron XP_011508507.1
XM_011510206.2 Intron XP_011508508.1
XM_011510207.2 Intron XP_011508509.1
XM_011510208.2 Intron XP_011508510.1
XM_011510209.2 Intron XP_011508511.1
XM_011510210.2 Intron XP_011508512.1
XM_017002966.1 Intron XP_016858455.1
XM_017002967.1 Intron XP_016858456.1
XM_017002968.1 Intron XP_016858457.1
XM_017002969.1 Intron XP_016858458.1
XM_017002970.1 Intron XP_016858459.1
XM_017002971.1 Intron XP_016858460.1
Gene
SMG7-AS1
Gene Name
SMG7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details