Product Details
- SNP ID
-
rs10908509
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
65
- Location
-
Chr.1:156580462 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTATGGAGGAGAGGCCCTGCATA[C/T]GCGGTTCCTTCATCTATTTATTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IQGAP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139012230,rs41405153] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IQGAP3
- Gene Name
- IQ motif containing GTPase activating protein 3
There are no transcripts associated with this gene.
- Gene
- TTC24
- Gene Name
- tetratricopeptide repeat domain 24
View Full Product Details