Product Details

SNP ID
rs13242635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55978963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAACTTTAAGACGAAAGGGGAGA[A/G]CTAAGCTTACCAGCTTTTAAGTTAA
Phenotype
MIM: 603004
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBAS PubMed Links

Gene Details

Gene
GBAS
Gene Name
glioblastoma amplified sequence
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202469.1 Intron NP_001189398.1
NM_001483.2 Intron NP_001474.1

View Full Product Details