Product Details

SNP ID
rs6528018
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:21394900 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATTGACTTTTCATAGGAAAAACAG[G/T]CAATTGTAAGAAACAAATGAACCAA
Phenotype
MIM: 300724
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNKSR2 PubMed Links

Gene Details

Gene
CNKSR2
Gene Name
connector enhancer of kinase suppressor of Ras 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168647.1 Intron NP_001162118.1
NM_001168648.1 Intron NP_001162119.1
NM_001168649.1 Intron NP_001162120.1
NM_014927.3 Intron NP_055742.2
XM_011545471.2 Intron XP_011543773.1
XM_011545472.2 Intron XP_011543774.1
XM_017029358.1 Intron XP_016884847.1

View Full Product Details