Product Details
- SNP ID
-
rs2247814
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:77089178 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTTTCAACAGAAAATGAGAAATATC[C/G]CCCGACGATTGGCTGAAAACATGCA
- Phenotype
-
MIM: 601504
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6516
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57684792] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6516
- Gene Name
- microRNA 6516
There are no transcripts associated with this gene.
- Gene
- SCARNA16
- Gene Name
- small Cajal body-specific RNA 16
There are no transcripts associated with this gene.
- Gene
- SEC14L1
- Gene Name
- SEC14 like lipid binding 1
- Gene
- SNHG20
- Gene Name
- small nucleolar RNA host gene 20
There are no transcripts associated with this gene.
View Full Product Details