Product Details

SNP ID
rs8071627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41872197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAGGCAACCTGGAGTGGGGGGA[A/T]CAAAGGCCAGGAGATGGTCGACAAG
Phenotype
MIM: 108728
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001096.2 Intron NP_001087.2
NM_001303274.1 Intron NP_001290203.1
NM_001303275.1 Intron NP_001290204.1
NM_198830.1 Intron NP_942127.1
XM_005257395.1 Intron XP_005257452.1
XM_017024688.1 Intron XP_016880177.1
Gene
KLHL11
Gene Name
kelch like family member 11
There are no transcripts associated with this gene.

View Full Product Details