Product Details

SNP ID
rs3744720
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:6579726 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATCCCTTCTCCAGTGTGACACAAA[C/T]GGCCTCGCCTCAGAGAGCCTTTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0753 PubMed Links

Gene Details

Gene
KIAA0753
Gene Name
KIAA0753
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014804.2 3082 UTR 3 NP_055619.2
XM_006721612.2 3082 UTR 3 XP_006721675.1
XM_011524090.2 3082 UTR 3 XP_011522392.1
XM_011524091.2 3082 UTR 3 XP_011522393.1
XM_011524095.2 3082 UTR 3 XP_011522397.1
XM_011524096.1 3082 Intron XP_011522398.1
XM_011524098.2 3082 UTR 3 XP_011522400.1
XM_011524099.2 3082 UTR 3 XP_011522401.1
XM_011524100.2 3082 UTR 3 XP_011522402.1
XM_017025455.1 3082 UTR 3 XP_016880944.1
XM_017025456.1 3082 UTR 3 XP_016880945.1
XM_017025457.1 3082 UTR 3 XP_016880946.1

View Full Product Details