Product Details

SNP ID
rs10511272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108590476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGAAAGATAAAAATGATTTAAC[C/T]AGTTGGGACATTCATGTAAAAATCT
Phenotype
MIM: 608672 MIM: 610643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DZIP3 PubMed Links

Gene Details

Gene
DZIP3
Gene Name
DAZ interacting zinc finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014648.3 Intron NP_055463.1
XM_005247914.3 Intron XP_005247971.1
XM_005247915.3 Intron XP_005247972.1
XM_005247916.3 Intron XP_005247973.1
XM_005247917.3 Intron XP_005247974.1
XM_005247918.3 Intron XP_005247975.1
XM_005247919.3 Intron XP_005247976.1
XM_017007540.1 Intron XP_016863029.1
Gene
KIAA1524
Gene Name
KIAA1524
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020890.2 Intron NP_065941.2
XM_006713716.3 Intron XP_006713779.1
XM_006713717.2 Intron XP_006713780.1
XM_011513056.1 Intron XP_011511358.1
XM_011513057.2 Intron XP_011511359.1

View Full Product Details