Product Details
- SNP ID
-
rs901963
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48340826 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTATTAAAGAGGGAGAGTAAAAGAA[A/G]TGTCAGAGTCCAGATTTATCACTGA
- Phenotype
-
MIM: 613906
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF641
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2634680] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF641
- Gene Name
- zinc finger protein 641
View Full Product Details