Product Details

SNP ID
rs2195951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9115009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTACACCCCAAAAGCTGTCCCA[T/C]AGAACAAGGAAAAAACAGAGAGGTG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
OR7G1 PubMed Links

Gene Details

Gene
OR7G1
Gene Name
olfactory receptor family 7 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005192.2 755 Missense Mutation TAT,TGT Y252C NP_001005192.2

View Full Product Details